missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORF73/HHV8 Antibody (LN53), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22767AF594
This item is not returnable.
View return policy
Description
ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Specifications
| ORF73/HHV8 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
| Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 935114 | |
| Store at 4°C in the dark. | |
| IgG2c |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| LN53 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction