missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORF73/HHV8 Rat anti-Virus, Clone: LN53, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22767-0.025MG
This item is not returnable.
View return policy
Description
ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoassay.
Specifications
| ORF73/HHV8 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunoassay | |
| HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody reacts with ORF-73 of human Herpes virus 8 (HHV8). HHV8 is the likely etiological agent of Kaposi's sarcoma (KS). HHV8 encodes a latent nuclear antigen (LNA) which is the product of the viral gene orf 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product which may be related to its oncogenic activity. | |
| Store at 4C. Do not freeze. | |
| IgG2c |
| Immunoassay, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| LN53 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| HHV8GP78 | |
| Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8. | |
| 0.025 mg | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 935114 | |
| Virus | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction