Learn More
New Pig Original DrainBlocker Drain Cover, 76cm x 76cm x 1.3cm
Brand:  New Pig PLRE245
Encompass_Preferred
Original DrainBlocker Drain Cover, 76cm x 76cm x 1.3cm
Product Code. 15721893
  • € 442.56 / Each
Only null left
Estimated Shipment: 06-02-2026