Learn More
B Braun™ Original Perfusor™ Lines
Product Code. 10669905
Change view
Click to view available options
Length (Metric):
50 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10669905 50 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10669905 Supplier B Braun™ Supplier No. 8255172
  • ONLINE EXCLUSIVE PRICE
    € 161.00
    € 1.53 / Pack of 1
    Save € 159.47
    99% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
For use with Perfusor automated infusion systems.
  • Luer lock fitting enables compatibility to all pump syringes
  • Pressure proof up to 2 bar
  • DEHP-free
Specifications
Diameter (Metric) Inner 1.5 mm
Length (Metric) 50 cm
Max. Pressure 2 bar
Quantity 100/Pk.
For Use With (Equipment) Perfusor Infusion Systems
Connection Type Luer Lock