Learn More
B Braun™ Original Perfusor™ Lines
Achieve accuracy in infusion therapy. B Braun™ Original Perfusor™ Lines are perfect for use with all syringe pumps used in infusion technology up to a pressure <2 bar.
Brand: B Braun™ 8255490
Additional Details : Weight : 0.70000kg
Description
- For all syringe pumps used in infusion technology up to a pressure <2 bar
- Available in different lengths and materials
- Minimal residual volume
- Luer lock approaches
Specifications
1.5 mm | |
250 cm | |
Syringe Pump |