Learn More
B Braun™ Original Perfusor™ Syringes with Aspiration Needle
Product Code. 10106073
Change view
Click to view available options
Volume (Metric):
20 mL
50 mL
Unit Size:
Each
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10106073 20 mL Pack of 100
10346534 50 mL Each
10294072 50 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10106073 Supplier B Braun™ Supplier No. 8728623
Use this 3-piece syringe for infusion pumps. B Braun™ Original Perfusor™ Syringes with Aspiration enable slow patient during infusion of drug solutions.
  • Polypropylene
  • High contrast graduation
  • Prmanent markings for readability
  • Graduation in ml increments
Specifications
Certifications/Compliance ISO 10993/DIN EN 30993, EN ISO 7886-1
Material Polypropylene
Product Type Syringe
Graduations 1 mL
Quantity 1/Ea.
Volume (Metric) 20 mL
For Use With (Equipment) Infusion Pump
Graduated Yes
Sterility Sterile