missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Orilanolimab Chimeric Recombinant Human Monoclonal Antibody

Description
Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Orilanolimab is a biosimilar that targets FCGRT.
Specifications
Specifications
| Antigen | Orilanolimab Chimeric |
| Applications | ELISA, SDS-Page |
| Classification | Recombinant Monoclonal |
| Concentration | 0.86 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.5 |
| Gene Alias | SYNT-001 |
| Host Species | Human |
| Immunogen | Human FCGRT. |
| Purification Method | Protein A/G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?