Learn More
Thermo Scientific™ Orion™ 2117XP Chloride Analyzer
Click to view available options
Includes:
Instrument Only Package
Protective Enclosure Package
Unit Size:
Each
Description
Detect chloride in real time with accurate and reliable results using the Thermo Scientific™ Orion™ 2117XP Chloride Analyzer. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification and peace of mind.
- Detection limit of 0.1ppm offers continuous chloride monitoring in wide variety of samples Protect against early breakthrough with sensitive, selective, reliable and verfiable measurements that provide early detection of chloride in RO and boiler systems
- Holds calibration up to 60 days between reagent changes Fast and highly accurate calibration has you back online in minutes
- Long life solid state sensor technology offers stability and high performance
- Simple operation and minimal maintenance required - no moving parts
- Repeatable and verifiable measurements at a glance using the large, user friendly display
2117XP Chloride Analyzer Ordering Information
- Annual Consumables Kit, Cat. No. 211750XP
- Reagent, Cat. No. 182011
- Diffusion Tubing, Cat. No. 181060
- Calibration Kit, Cat. No. 2117XPCAL
- Calibration Standards, Cat. No. 211740
- Chloride Electrode, Cat. No. 100025
- Reference Electrode, Cat. No. 100057
- Reference Electrode Filling Solution, Cat. No. 100057F4
Specifications
Specifications
| Accuracy | ±10% or 0.1 ppm, whichever is greater |
| Accuracy (mV) | ± (0.5 mV + 0.1%) |
| Accuracy (Temperature) | ±0.5°C |
| Alarm Output | 3 |
| Analog Outputs | 2, Galvanically Isolated |
| Calibration | Double Known Addition |
| Case Material | Valox 364 |
| Certifications/Compliance | CE, CSA, UL, FCC Class A limits |
| Channels | 1 |
| Communication Details | Modbus with Additional Module |
| Show More |