Learn More
Thermo Scientific™ Orion™ 2117XP Chloride Analyzer
Product Code. p-4529544
Click to view available options
Includes:
Instrument Only Package
Protective Enclosure Package
Unit Size:
Each
Product Code. 16582378
Brand: Thermo Scientific™ 2117XP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Detect chloride in real time with accurate and reliable results using the Thermo Scientific™ Orion™ 2117XP Chloride Analyzer.
Detect chloride in real time with accurate and reliable results using the Thermo Scientific™ Orion™ 2117XP Chloride Analyzer. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification and peace of mind.
  • Detection limit of 0.1ppm offers continuous chloride monitoring in wide variety of samples Protect against early breakthrough with sensitive, selective, reliable and verfiable measurements that provide early detection of chloride in RO and boiler systems
  • Holds calibration up to 60 days between reagent changes Fast and highly accurate calibration has you back online in minutes
  • Long life solid state sensor technology offers stability and high performance
  • Simple operation and minimal maintenance required - no moving parts
  • Repeatable and verifiable measurements at a glance using the large, user friendly display
2117XP Chloride Analyzer Ordering Information
  • Annual Consumables Kit, Cat. No. 211750XP
  • Reagent, Cat. No. 182011
  • Diffusion Tubing, Cat. No. 181060
  • Calibration Kit, Cat. No. 2117XPCAL
  • Calibration Standards, Cat. No. 211740
  • Chloride Electrode, Cat. No. 100025
  • Reference Electrode, Cat. No. 100057
  • Reference Electrode Filling Solution, Cat. No. 100057F4
Specifications
Accuracy ±10% or 0.1 ppm, whichever is greater
Accuracy (mV) ± (0.5 mV + 0.1%)
Accuracy (Temperature) ±0.5°C
Alarm Output 3
Analog Outputs 2, Galvanically Isolated
Calibration Double Known Addition
Case Material Valox 364
Certifications/Compliance CE, CSA, UL, FCC Class A limits
Channels 1
Communication Details Modbus with Additional Module
Current Loops 2
Description 2117XP Chloride Analyzer
Detection Limits 0.1ppm
Temperature (Metric) Operating 5°C to 35°C
Type Chloride Analyzer
Display Type Custom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure Rating Enclosure not included
Flow Rate 40mL/min.
Includes Instrument Only Package
Inlet Pressure 8 to 100psig
Keypad Type Membrane
Max. Relative Humidity 5 to 95%, non-condensing
Measurement Ranges 0 to 10ppm
Mounting Options Wall mount
Outputs 0 to 20 mA; 4 to 20 mA
Relay Details 3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Response Time 95% within 2 minutes of injecting a standard solution
Sample Flow Rate 40 mL/min nominal set by pressure regulator
Sample Inlet 1/4 in. NPTF tube fitting
Sample Streams 1
Dimensions (L x W x H) 11 x 17 x 26 in. (27 x 45 x 65cm)
Length (English) 11 in.
Length (Metric) 27 cm
Width (English) 17 in.
Width (Metric) 45 cm
Height (English) 26 in.
Height (Metric) 65 cm
Voltage 85/132 VAC, 170/264 VAC
Amperage 5A
Frequency 0 to 60 Hz
Channel 1 Chloride
Channel 2 Optional ph/ORP or Conductivity