Learn More
Thermo Scientific™ Orion™ AquaPro™ Double Junction Combination pH Electrodes
Description
Thermo Scientific™ Orion™ AquaPro™ Low Maintenance pH Electrodes provide fast response and extended life with the additional advantages of a double junction design. Their clog resistant, open junctions allow for use in difficult samples, while making post-use cleaning, simple. They are available in a variety of different designs, with the standard types being the 9104APWP & the 9156APWP. These have glass and epoxy bodies respectively and are designed for general lab use. The 9135APWP has a uniquely designed, flat sensing bulb that allows for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates. The 9107APMD contains a built-in temperature sensor that accurately measures pH and temperature with just one electrode, so combines the fast response and open junction design of the Orion™ AquaPro™ Electrode with a built-in ATC.
Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.
- Measure 0 to 14pH
- Temperature range: 0° to 60°C (0° to 140°F)
- Choice of connectors with 1m (3ft.) cable
Specifications
Specifications
| Description | Combination semi-micro pH electrode, Glass Body pH |
| Connector Types | BNC (Waterproof) |
| Junction | Open |
| Material | Glass Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 60°C |
| Type | Combination pH Electrode |
| For Use With (Application) | Small sample sizes, test tubes |
| Internal Reference | Ag/AgCl Double Junction |
| pH Relative Accuracy | ±0.02 |
| Show More |