Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Cadmium Combination Ion Selective Electrode
Product Code. 10489102
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10489102 BNC (Waterproof) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10489102 Supplier Thermo Scientific™ Supplier No. 9648BNWP
  • ONLINE EXCLUSIVE PRICE
    € 1055.00
    € 842.40 / Pack of 1
    Save € 212.60
    20% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure cadmium ions in aqueous solutions quickly, simply, accurately and economically.
Measure cadmium ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Cadmium Electrodes.
Features:
  • 9648BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Solutions required:
  • ISA
    • 940011 ISA for cadmium measurements,475 mL bottle
  • Electrode fill solution
    • 900061 Optimum Results A electrode filling solution, 5 x 60 mL bottles
Items included with:
  • 900061 Optimum Results A electrode filling solution. 60 mL 1 bottle
  • 948201 Polishing strip. Qty 1.
Warranty period
  • 12 months from date of purchase
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC (Waterproof)
Description Model 9448BN; Orion Cadmium Combination
Junction Sure-Flow Junction
For Use With (Equipment) ISE Meter
Type Combination ISE