Learn More
Thermo Scientific™ Orion™ Swing Arm Electrode Stand
Product Code. 15977115
Click to view available options
:
Swing Arm and Holder
Product Code. 15977115
Brand: Thermo Scientific™ 090043
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Quickly move electrodes between samples with the Thermo Scientific™ Orion™ Swing Arm Electrode Stand, featuring 360 degrees of motion and cable management system
Quickly move electrodes between samples with the Thermo Scientific™ Orion™ Swing Arm Electrode Stand, featuring 360 degrees of motion and cable management system for neatness and safety. Includes weighted base, swing arm and electrode holder that accepts up to four sensors and stirrer probe.
  • Holds up to five electrodes with 12mm diameters; or four electrodes and one stirrer probe
  • Aluminum construction for stability and strength
  • 360° motion allows electrodes to be moved vertically, horizontally and front-to-back positioning into beakers or sample containers
  • Head holds three electrodes vertically (12mm max. diameter)
  • Two additional slots hold electrodes angled 10° (12mm max. diameter)
  • 5.5″ diameter weighted base (140mm) with cushioning on the bottom to protect table surfaces
TRUSTED_SUSTAINABILITY
Specifications
Description Orion swing arm stand, electrode holder and base
Type Electrode Stand/Holder
Includes Standard electrode holder (five 12mm diameter holders), metal stand with cable management system and weighted base
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.