missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORM1 Monoclonal Antibody (C1)

Product Code. 17886412
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17886412 200 μL 200µL
17634096 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17886412 Supplier Invitrogen™ Supplier No. MA541543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ORM1 Monoclonal Antibody for Western Blot, IHC (P)

ORM1 is an acute phase plasma protein synthesized by the liver. The protein is believed to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, also regulates the extravasation of the cells during infection and inflammation. ORM1 is a major acute phase reactant, both in human and in rat. In human plasma, ORM1 (44 kDa) is found at levels of 0.5-1.4 mg/mL. Its concentration in blood plasma is elevated during acute inflammation, such as trauma, malignancies, myocardial infarction, rheumatoid arthritis, bacterial infections, after major surgery, in cases of chronic pain etc, and can be used for the diagnosis of inflammatory conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ORM1
Gene Accession No. P02763, P02764
Gene Alias AGP; AGP 1; Agp1; Agp-1; Agp-2; AGP-A; Agpa1; alpha-1 acid glycoprotein; alpha-1-acid glycoprotein; Alpha-1-acid glycoprotein 1; alpha-1-acid glycoprotein-2; bM340H1.2 (orosomucoid 1); epididymis secretory sperm binding protein Li 153w; HEL-S-153w; OMD; OMD 1; Orm; Orm1; Orm-1; Orosomucoid; orosomucoid 1; orosomucoid-1
Gene Symbols ORM1
Host Species Mouse
Immunogen Recombinant protein Alpha-1-Acid Glycoprotein. The antigen corresponds to amino acid range 19-201 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24614, 5004
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.