missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORM1 Monoclonal Antibody (D6)

Mouse Monoclonal Antibody
€ 156.00 - € 442.00
Specifications
| Antigen | ORM1 |
|---|---|
| Clone | D6 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17868802
![]() |
Invitrogen™
MA541544 |
200 μL |
€ 442.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17624096
|
Invitrogen™
MA543638 |
20 μL |
€ 156.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ORM1 Monoclonal Antibody for Western Blot, IHC (P)
ORM1 is an acute phase plasma protein synthesized by the liver. The protein is believed to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, also regulates the extravasation of the cells during infection and inflammation. ORM1 is a major acute phase reactant, both in human and in rat. In human plasma, ORM1 (44 kDa) is found at levels of 0.5-1.4 mg/mL. Its concentration in blood plasma is elevated during acute inflammation, such as trauma, malignancies, myocardial infarction, rheumatoid arthritis, bacterial infections, after major surgery, in cases of chronic pain etc, and can be used for the diagnosis of inflammatory conditions.Specifications
| ORM1 | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P02763 | |
| 5004 | |
| Recombinant protein Alpha-1-Acid Glycoprotein. The antigen corresponds to amino acid range 19-201 of the target protein. | |
| Antibody |
| D6 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| ORM1 | |
| AGP; AGP 1; Agp1; Agp-1; Agp-2; AGP-A; Agpa1; alpha-1 acid glycoprotein; alpha-1-acid glycoprotein; Alpha-1-acid glycoprotein 1; alpha-1-acid glycoprotein-2; bM340H1.2 (orosomucoid 1); epididymis secretory sperm binding protein Li 153w; HEL-S-153w; OMD; OMD 1; Orm; Orm1; Orm-1; Orosomucoid; orosomucoid 1; orosomucoid-1 | |
| ORM1 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title