missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORMDL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76577-0.025mg
This item is not returnable.
View return policy
Description
ORMDL1 Polyclonal specifically detects ORMDL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ORMDL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| adoplin-1, DKFZp686G141, FLJ44364, ORM1 (S. cerevisiae)-like 1, ORM1-like 1 (S. cerevisiae), ORM1-like protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 94101 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_057551 | |
| ORMDL1 | |
| Antibody was raised against a 16 amino acid synthetic peptide of human ORMDL1. The immunogen is located within amino acids 50 - 100 of ORMDL1. Amino Acid Squence: VKGTPFETPDQGKARL | |
| 0.025 mg | |
| Primary | |
| ORMDL1 antibody may cross-react with other ORMDL protein family members. | |
| Store at 4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction