Learn More
ORMDL3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95148-0.1ml
This item is not returnable.
View return policy
Description
ORMDL3 Polyclonal antibody specifically detects ORMDL3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ORMDL3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ORM1 (S. cerevisiae)-like 3, ORM1-like 3 (S. cerevisiae), ORM1-like protein 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ORMDL3 (NP_644809.1). MNVGTAHSEVNPNTRVMNSRGIWLSYVLAIGLLHIVLLSIPFVSVPVVWTLTNLIHNMGMYIFLHTVKGTPFETPDQGKARLLTHWEQMDYGVQFTASRK | |
| 0.1 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 94103 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |