missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Carbamoyltransferase Antibody (OTI8A1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73149B
This item is not returnable.
View return policy
Description
Ornithine Carbamoyltransferase Monoclonal specifically detects Ornithine Carbamoyltransferase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Ornithine Carbamoyltransferase | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| OTC | |
| Recombinant protein fragment corresponding to amino acids 33-354 of human Ornithine Carbamoyltransferase (NP_000522)produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI8A1 | |
| Western Blot | |
| EC 2.1.3, EC 2.1.3.3, MGC129967, MGC129968, MGC138856, OCTD, ornithine carbamoyltransferase, ornithine carbamoyltransferase, mitochondrial, Ornithine transcarbamylase, OTCase | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 5009.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction