Learn More
Ornithine Decarboxylase Antibody (ODC1/487), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47837PE
Code produit. 18621666
  • € 544.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Ornithine Decarboxylase
Monoclonal
PE
ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1
Recombinant human Ornithine Decarboxylase protein (Uniprot: P11926)
0.1 mL
Cell Cycle and Replication
4953
Store at 4°C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
ODC1/487