missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Antibody (rODC1/485), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08785APC
This item is not returnable.
View return policy
Description
Ornithine Decarboxylase Monoclonal antibody specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Ornithine Decarboxylase | |
| Monoclonal | |
| APC | |
| ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
| Recombinant full-length human Ornithine Decarboxylase protein (Uniprot: P11926) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 4953 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rODC1/485 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction