missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ornithine Decarboxylase Mouse, Clone: ODC1/485, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32887-0.1MG
This item is not returnable.
View return policy
Description
Ornithine Decarboxylase Monoclonal specifically detects Ornithine Decarboxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Ornithine Decarboxylase | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| ODCEC 4.1.1.17, ornithine decarboxylase, ornithine decarboxylase 1 | |
| Mouse | |
| 53 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 4953 | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| ODC1/485 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page, Immunofluorescence | |
| P11926 | |
| ODC1 | |
| Recombinant human ODC-1 protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa. | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction