Learn More
ORP1 Antibody (S02-2E2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19727-100ul
 View more versions of this product
Product Code. 18668203
  • € 645.00 / 100µL
Description
ORP1 Monoclonal antibody specifically detects ORP1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ORP1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S02-2E2
Unconjugated
FLJ10217, ORP1oxysterol-binding protein-related protein 1, ORP-1oxysterol-binding protein-related protein 1 variant 1, OSBP8, OSBPL1, OSBPL1B, OSBP-related protein 1, oxysterol binding protein-like 1A, oxysterol binding protein-like 1B, oxysterol-binding protein-related protein 1 variant 2
A synthetic peptide of human ORP1 (Uniprot # Q9BXW6)
100 μg
Lipid and Metabolism
114876
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.