missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17109053
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17109053 100 μL 100µL
17149093 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17109053 Supplier Invitrogen™ Supplier No. PA5111129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VPKNSLQQSR EDWLEAIEEH SAYSTHYCSQ DQLTDEEEED TVSAADLKKS LEKAQSCQQR LDREISNFLK MIKECDMAKE MLPSFLQKVE VVSEA Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 82%.

This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OSBPL1A
Gene Accession No. Q9BXW6
Gene Alias DCDC4A; FLJ10217; FLJ50293; FLJ55454; FLJ79410; G430090F17Rik; Gm753; ORP1; ORP-1; Orp1a; Orp1l; OSBP8; OSBPL1; OSBPL1A; OSBPL1B; OSBP-related protein 1; oxysterol binding protein like 1A; oxysterol binding protein-like 1A; oxysterol binding protein-like 1B; oxysterol-binding protein-related protein 1; oxysterol-binding protein-related protein 1 variant 1; oxysterol-binding protein-related protein 1 variant 2
Gene Symbols OSBPL1A
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ORP1. Recombinant protein control fragment (Product #RP-98278).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114876
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.