Learn More
Invitrogen™ ORP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140154
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ESLHRIPSAP VIPTHQASVT TERPKKGKRT SRMWCTQSFA KDDTIGRVGR LHGSVPNLSR YLESRDSSGT RGLPPTDYAH LQRSFWALAQ KVHSSL Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.
Specifications
| ORP7 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL7 | |
| ORP7; ORP-7; OSBPL7; OSBP-related protein 7; oxysterol binding protein like 7; oxysterol binding protein-like 7; oxysterol-binding protein-related protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114881 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZF2 | |
| OSBPL7 | |
| Recombinant protein corresponding to Human OSBPL7. Recombinant protein control fragment (Product #RP-109380). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.