missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP8 Antibody (4H6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00114882-M02
This item is not returnable.
View return policy
Description
ORP8 Monoclonal antibody specifically detects ORP8 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| ORP8 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| MGC126578, MGC133203, MST120, MSTP120, ORP-8, ORP8KIAA1451, OSBP10DKFZp686A11164, OSBP-related protein 8, oxysterol binding protein-like 8, oxysterol-binding protein-related protein 8 | |
| OSBPL8 (NP_065892.1, 244 a.a. ∽ 346 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. IIRATSESDGRCWMDALELALKCSSLLKRTMIREGKEHDLSVSSDSTHVTFYGLLRANNLHSGDNFQLNDSEIERQHFKDQDMYSDKSDKENDQEHDESDNEV | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Sandwich ELISA | |
| 4H6 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_065892 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 114882 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction