missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35792-100ul
This item is not returnable.
View return policy
Description
ORP8 Polyclonal antibody specifically detects ORP8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ORP8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| MGC126578, MGC133203, MST120, MSTP120, ORP-8, ORP8KIAA1451, OSBP10DKFZp686A11164, OSBP-related protein 8, oxysterol binding protein-like 8, oxysterol-binding protein-related protein 8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human ORP8 (NP_065892.1).,, Sequence:, MEGGLADGEPDRTSLLGDSKDVLGPSTVVANSDESQLLTPGKMSQRQGKEAYPTPTKDLHQPSLSPASPHSQGFERGKEDISQNKDESSLSMSKSKSESKLYNGSEKDSS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 114882 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction