missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OS9 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97288AF750
This item is not returnable.
View return policy
Description
OS9 Polyclonal antibody specifically detects OS9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OS9 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| Amplified in osteosarcoma 9, endoplasmic reticulum lectin 2, ERLEC2, erlectin 2, OS-9, osteosarcoma amplified 9, endoplasmic reticulum associated protein, osteosarcoma amplified 9, endoplasmic reticulum lectin, protein OS-9 | |
| Produced in rabbits immunized with E. coli-derived Human OS9 fragment. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Hypoxia, Immunology | |
| 10956 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction