missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBP1 Antibody (OTI1F2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-00935
This item is not returnable.
View return policy
Description
OSBP1 Monoclonal specifically detects OSBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| OSBP1 | |
| Monoclonal | |
| 0.33 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| OSBP | |
| Full length human recombinant protein of human OSBP (NP_002547) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1F2 | |
| Unconjugated | |
| OSBP1oxysterol-binding protein 1, oxysterol binding protein | |
| Mouse | |
| 89.2 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5007 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction