missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBP1 Antibody (OTI1F2), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73152PE
This item is not returnable.
View return policy
Description
OSBP1 Monoclonal specifically detects OSBP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| OSBP1 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| OSBP | |
| Full length recombinant protein of human OSBP1 (NP_002547) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1F2 | |
| Flow Cytometry | |
| OSBP1oxysterol-binding protein 1, oxysterol binding protein | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 5007.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction