missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL11 Monoclonal Antibody (OTI2F6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501938S
This item is not returnable.
View return policy
Description
OSBPL11 Monoclonal antibody specifically detects OSBPL11 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| OSBPL11 | |
| Monoclonal | |
| 0.61 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UJM8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54363 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI2F6 | |
| Unconjugated | |
| HAO1 | |
| 9430097N02Rik; AI132306; AU019795; FLJ13012; FLJ13164; ORP11; ORP-11; OSBP12; OSBPL11; OSBP-related protein 11; oxysterol binding protein like 11; oxysterol binding protein-like 11; oxysterol-binding protein-related protein 11; TCCCIA00292; testicular tissue protein Li 132 | |
| Full length human recombinant protein of human HAO1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction