missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35419-20ul
This item is not returnable.
View return policy
Description
OSBPL2 Polyclonal antibody specifically detects OSBPL2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| OSBPL2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| KIAA0772FLJ20223, ORP-2MGC4307, ORP2MGC8342, OSBP-related protein 2, oxysterol binding protein-like 2, oxysterol-binding protein-like 2, oxysterol-binding protein-related protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 291-480 of human OSBPL2 (NP_653081.1).,, Sequence:, NKKKLFMIYGKWTECLWGIDPVSYESFKKQERRGDHLRKAKLDEDSGKADSDVADDVPVAQETVQVIPGSKLLWRINTRPPNSAQMYNFTSFTVSLNELETGMEKTLPPTDCRLRPDIRGMENGNMDLASQEKERLEEKQREARRERAKEEAEWQTRWFYPGNNPYTGTPDWLYAGDYFERNFSDCPDIY | |
| 20 μL | |
| Lipid and Metabolism | |
| 9885 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction