missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31065
This item is not returnable.
View return policy
Description
OSBPL2 Polyclonal specifically detects OSBPL2 in Human samples. It is validated for Western Blot.
Specifications
| OSBPL2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| OSBPL2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human OSBPL2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (85%) and Xenopus tropicalis (85%). Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| KIAA0772FLJ20223, ORP-2MGC4307, ORP2MGC8342, OSBP-related protein 2, oxysterol binding protein-like 2, oxysterol-binding protein-like 2, oxysterol-binding protein-related protein 2 | |
| Rabbit | |
| 55 kDa | |
| 100 μL | |
| Lipid and Metabolism | |
| 9885 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction