Learn More
OSBPL2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97886PE
This item is not returnable.
View return policy
Description
OSBPL2 Polyclonal antibody specifically detects OSBPL2 in Human samples. It is validated for Immunohistochemistry (Paraffin)