missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-77443
This item is not returnable.
View return policy
Description
OSBPL3 Polyclonal antibody specifically detects OSBPL3 in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence
Specifications
| OSBPL3 | |
| Polyclonal | |
| Flow Cytometry 10 μg/mL, Peptide ELISA Detection limit 1:32000, Immunofluorescence 10 μg/mL | |
| DKFZp667P1518, KIAA0704ORP3ORP-3OSBP3, MGC21526, OSBP-related protein 3, oxysterol binding protein-like 3, oxysterol-binding protein 3, oxysterol-binding protein-related protein 3 | |
| Peptide with sequence MSDEKNLGVSQKL-C corresponding to N-Terminus according to NP_056365.1, NP_663160.1, NP_663161.1, NP_663162.1. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Flow Cytometry, Peptide ELISA, Immunofluorescence | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| 26031 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction