missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSCAR Antibody (MM0502-6S6), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11835AF750
This item is not returnable.
View return policy
Description
OSCAR Monoclonal specifically detects OSCAR in Human samples. It is validated for Western Blot.
Specifications
| OSCAR | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| OSCAR | |
| Human recombinant OSCAR (extracellular domain) | |
| Immunology | |
| 126014 | |
| Store at 4°C in the dark. |
| Western Blot | |
| MM0502-6S6 | |
| Western Blot | |
| hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction