missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSCAR Antibody (RM0300-11E34), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11836AF700
This item is not returnable.
View return policy
Description
OSCAR Monoclonal specifically detects OSCAR in Mouse samples. It is validated for Western Blot.
Specifications
| OSCAR | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| OSCAR | |
| Mouse recombinant OSCAR (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0300-11E34 | |
| Western Blot | |
| hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
| Rat | |
| Protein A or G purified | |
| Immunology | |
| 126014 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction