missing translation for 'onlineSavingsMsg'
Learn More

OSGEP Antibody (OTI9E3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73163B

Product Code. 18111976

  • € 785.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OSGEP Monoclonal specifically detects OSGEP in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OSGEP
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
OSGEP
Recombinant protein fragment corresponding to amino acids 1-335 of human OSGEP (NP_060277) produced in E.coli.
0.1 mL
55644.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry
OTI9E3
Western Blot, Flow Cytometry, Immunohistochemistry
EC 3.4.24.57, GCPL1FLJ20411, hOSGEP, KAE1, OSGEP1, O-sialoglycoprotein endopeptidase, probable O-sialoglycoprotein endopeptidase, PRSMG1, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP
Mouse
Affinity Purified
Primary
Human, Mouse, Rat, Canine
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.