missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGEP Antibody (OTI9E3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73163MFV610
This item is not returnable.
View return policy
Description
OSGEP Monoclonal antibody specifically detects OSGEP in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
Specifications
| OSGEP | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
| Flow Cytometry | |
| OTI9E3 | |
| Flow Cytometry | |
| EC 3.4.24.57, GCPL1FLJ20411, hOSGEP, KAE1, OSGEP1, O-sialoglycoprotein endopeptidase, probable O-sialoglycoprotein endopeptidase, PRSMG1, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP | |
| Human recombinant protein fragment corresponding to amino acids 1-335 of human OSGEP(NP_060277) produced in E.coli. | |
| 0.1 mL | |
| Cell Biology | |
| 55644 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction