missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGIN2 Antibody (OTI7D12), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73164B
This item is not returnable.
View return policy
Description
OSGIN2 Monoclonal specifically detects OSGIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| OSGIN2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| OSGIN2 | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human OSGIN2 (NP_004328) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI7D12 | |
| Western Blot, Immunohistochemistry | |
| hT41C8orf1chromosome 8 open reading frame 1, oxidative stress induced growth inhibitor family member 2, oxidative stress-induced growth inhibitor 2 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 734.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction