missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGIN2 Monoclonal Antibody (OTI7D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808943S
This item is not returnable.
View return policy
Description
OSGIN2 Monoclonal antibody specifically detects OSGIN2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| OSGIN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP98 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 58529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7D12 | |
| Unconjugated | |
| MYOZ1 | |
| BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323 | |
| Human recombinant protein fragment corresponding to amino acids 106-299 of human MYOZ1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction