missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSGIN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565627
This item is not returnable.
View return policy
Description
Immunogen sequence: EGSMLTISFGS WMELPGLKFK DWVSSKRRSL KGDRVMPEEI ARYYKHYVKV MGLQKNFREN TYITSVSRLY RDQDDDDIQD RDISTKHLQ Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
May be involved in meiosis or the maturation of germ cells.
Specifications
| OSGIN2 | |
| Polyclonal | |
| Unconjugated | |
| OSGIN2 | |
| BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 734 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y236 | |
| OSGIN2 | |
| Recombinant Human OSGIN2. Recombinant protein control fragment (Product #RP-106294). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction