missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress OSM Protein, Human (209a.a)

Product Code. 16489246
Encompass_Preferred
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16489246

Brand: MedChemExpress HYP7052A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OSM Protein, Human (209a.a) is a polypeptide chain containing 209 amino acids. Oncostatin M is a cytokine belonging to the IL-6 family that has multiple functions in hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, heart remodeling, nociception, inflammation and metabolism.

Specifications

Content And Storage Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage.
For Use With (Application) Neuroscience-Neuromodulation
Quantity 100 μg
Shipping Condition Room temperature
Molecular Weight (g/mol) Approximately 23.8 kDa
Synonym rHuOSM∣ OSM
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.