Learn More
OSMR beta Antibody (469221), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4389MFV500
Product Code. 18504237
  • € 674.00 / 0.10mL
Description
OSMR beta Monoclonal antibody specifically detects OSMR beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OSMR beta
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Primary
Human
IgG1
Western Blot, Flow Cytometry, ELISA, CyTOF, KnockDown
469221
Western Blot, Flow Cytometry, ELISA, CyTOF-ready, Knockout Validated
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Mouse myeloma cell line NS0-derived recombinant human OSM R beta , Glu28-Ser739, Accession # Q99650
RUO
9180
Store at 4C in the dark.