missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ OSMR beta Antibody (vixarelimab) - IgG4SP, Novus Biologicals™

Human Monoclonal Antibody

€ 213.00 - € 920.00

Specifications

Antigen OSMR beta
Clone vixarelimab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30044229
View Documents
Novus Biologicals™
NBP3-28875-100ug
100 μg
ONLINE EXCLUSIVE PRICE
€ 349.00
€ 329.70 / 100µg
Save € 19.30
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046009
View Documents
Novus Biologicals™
NBP3-28875-50ug
50 μg
€ 213.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044750
View Documents
Novus Biologicals™
NBP3-28875-1mg
1 mg
€ 920.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OSMR beta Monoclonal antibody specifically detects OSMR beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

OSMR beta
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Human
Q99650
9180
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
vixarelimab
Monoclonal
Purified
RUO
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
OSMR
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.