missing translation for 'onlineSavingsMsg'
Learn More

OSR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-30669-25ul

 View more versions of this product

Product Code. 18476781

  • € 280.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OSR2 Polyclonal specifically detects OSR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OSR2
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunodiffusion
Q8N2R0
OSR2
This antibody was developed against a recombinant protein corresponding to amino acids: TMHMNHWTLGYPNVHEITRSTITEMAAAQGLVDARFPFPALPFTTHLFHPKQGAIAHVLPALHKDRPRFDFANL
25 μL
DNA replication Transcription Translation and Splicing
116039
Human
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunodiffusion
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
odd-skipped related 2 (Drosophila), protein odd-skipped-related 2
Rabbit
Affinity Purified
RUO
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only