missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteoadherin/OSAD/OMD Antibody (014), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89674CL1
This item is not returnable.
View return policy
Description
Osteoadherin/OSAD/OMD Monoclonal antibody specifically detects Osteoadherin/OSAD/OMD in Human samples. It is validated for ELISA
Specifications
| Osteoadherin/OSAD/OMD | |
| Monoclonal | |
| CoraFluor 1 | |
| Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2Costeoadherin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Osteoadherin/OSAD/OMD (Accession#: NP_005005.1; Met1-Glu421). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 014 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4958 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction