missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteocalcin Antibody (2H9F11F8), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62801AF488
This item is not returnable.
View return policy
Description
Osteocalcin Monoclonal specifically detects Osteocalcin in Human, Rat, Porcine, Bovine, Rabbit, Mouse (Negative) samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| Osteocalcin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BGLAP | |
| Recombinant human osteocalcin protein | |
| 0.1 mL | |
| Extracellular Matrix, Stem Cell Markers | |
| 632 | |
| Human, Rat, Pig, Bovine, Rabbit, Mouse (Negative) | |
| Purified |
| ELISA, Immunohistochemistry | |
| 2H9F11F8 | |
| ELISA, Immunohistochemistry | |
| BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This product recognizes intact human and bovine osteocalcin in a “2 site” assay and reacts with osteocalcin fragments corresponding to amino acids 15-31 and 20-43 of the native molecule. It does not recognize fragments representing amino acids 1-19 or 7-19. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction