missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteocalcin Antibody (582014), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB113272A
This item is not returnable.
View return policy
Description
Osteocalcin Monoclonal antibody specifically detects Osteocalcin in Human samples. It is validated for Flow Cytometry
Specifications
| Osteocalcin | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 582014 | |
| Intracellular Staining by Flow Cytometry | |
| BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin | |
| Osteocalcin/KLH containing peptide, Accession # P02818 | |
| 0.1 mL | |
| Extracellular Matrix, Stem Cell Markers | |
| 632 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction