missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin/OPN Antibody (OTI5E4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73165AF700
This item is not returnable.
View return policy
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Osteopontin/OPN | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPP1 | |
| Recombinant protein fragment corresponding to amino acids 17-314 of human Osteopontin/OPN (NP_001035147) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5E4 | |
| Western Blot, Immunohistochemistry | |
| BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Extracellular Matrix, Hypoxia | |
| 6696.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction