missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Osteopontin/OPN Antibody Blocking Peptide

Product Code. 18243153 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18243153

Brand: Novus Biologicals™ NB6001043PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from human Osteopontin/OPN. Source: Synthetic The Osteopontin/OPN Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number p10451
Gene ID (Entrez) 6696
Species Human
Protein Family Osteopontin/OPN
Purity >95%
Concentration 1.0 mg/mL
Content And Storage Store at -20 C short term. Aliquot and store (at -80 C long term. Avoid freeze-thaw cycles.)
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
For Use With (Application) Antibody Competition
Gene Symbol SPP1
Quantity 0.05 mg
Regulatory Status RUO - research use only
Research Category Cellular Markers
Source Synthetic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.