missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteoprotegerin/TNFRSF11B Antibody (155321), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8052MFV610
This item is not returnable.
View return policy
Description
Osteoprotegerin/TNFRSF11B Monoclonal antibody specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Osteoprotegerin/TNFRSF11B | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cell Biology | |
| 4982 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 155321 | |
| Flow Cytometry, CyTOF-ready | |
| OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| Mouse myeloma cell line, NS0-derived human Osteoprotegerin/TNFRSF11B, Glu22-Leu401, Accession # AAB53709.1 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction